Learn More
Medchemexpress LLC HY-117286 10mg Medchemexpress, (S)-JQ-35 CAS:1349719-98-7 Purity:>98%
SDP
Catalog No. 501969464
Encompass_Preferred
Medchemexpress, HY-117286 10mg (S)-JQ-35 CAS:1349719-98-7 (S)-JQ-35 (TEN-010) is an inhibitor of the Bromodomain and Extra-Terminal (BET) family bromodomain-containing proteins with potential antineoplastic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9464 Supplier Medchemexpress LLC Supplier No. HY11728610MG
Medchemexpress, HY-117286 10mg (S)-JQ-35 CAS:1349719-98-7 (S)-JQ-35 (TEN-010) is an inhibitor of the Bromodomain and Extra-Terminal (BET) family bromodomain-containing proteins with potential antineoplastic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.