Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC S-JQ-35 10mM 1mL | 1349719-98-7 | 1 ML
SDP

Catalog No. 5000226792
Encompass_Preferred

(S)-JQ-35 (TEN-010) is a selective inhibitor of bromodomain and extra-terminal (BET) proteins, active against BRD2, BRD3, BRD4, and BRDT. Supplied as a 10 mM solution in DMSO (1 mL), it is intended for biochemical and cell-based research applications.

  • Selective BET bromodomain inhibition for BRD2, BRD3, BRD4, and BRDT studies.
  • Supplied as a 10 mM solution in DMSO, ready for in vitro assays.
  • High purity (99.46%), suitable for research applications.
  • Physical form: solid stock material; solution appearance light yellow to pink.
  • Typical applications: biochemical assays, cell-based assays, and screening studies.

Catalog No. 50-002-26792 Supplier Medchemexpress LLC Supplier No. HY11728610MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-JQ-35 (TEN-010) is a selective inhibitor of bromodomain and extra-terminal (BET) proteins, active against BRD2, BRD3, BRD4, and BRDT. Supplied as a 10 mM solution in DMSO (1 mL), it is intended for biochemical and cell-based research applications.

  • Selective BET bromodomain inhibition for BRD2, BRD3, BRD4, and BRDT studies.
  • Supplied as a 10 mM solution in DMSO, ready for in vitro assays.
  • High purity (99.46%), suitable for research applications.
  • Physical form: solid stock material; solution appearance light yellow to pink.
  • Typical applications: biochemical assays, cell-based assays, and screening studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.