Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-Ketorolac | 66635-92-5 | 99.3% | 25 MG
SDP

Catalog No. 5000450757
Encompass_Preferred

(S)-Ketorolac is a nonsteroidal anti-inflammatory agent that exhibits potent COX1 and COX2 enzyme inhibition.

  • Target: COX1 and COX2
  • Molecular formula: C15H13NO3
  • Molecular weight: 255.27
  • Appearance: White to off-white solid
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 2 years, -20°C for 1 year
  • Solubility in DMSO: 200 mg/mL

Catalog No. 50-004-50757 Supplier Medchemexpress LLC Supplier No. 25MGHYB0580A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-Ketorolac is a nonsteroidal anti-inflammatory agent that exhibits potent COX1 and COX2 enzyme inhibition.

  • Target: COX1 and COX2
  • Molecular formula: C15H13NO3
  • Molecular weight: 255.27
  • Appearance: White to off-white solid
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 2 years, -20°C for 1 year
  • Solubility in DMSO: 200 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.