Learn More
Thermo Scientific Thermostat Kit for 4 Channel Pump Kit
Supplier:  Thermo Scientific 842315550251
Encompass
repair kit for refrigerator contains tubing, fittings, seals, valves, filter driers, and compressor start components to service leaks, restrictions, and update thermo scientific refrigeration system without replacing entire system
Catalog No. NC0639219