Learn More
STERIS™ S-Klenz Scale Remover
Catalog No. 19999648 Shop All STERIS Corporation Products
Change view
Click to view available options
Volume (English):
1 gal.
Catalog No. Volume (English)
19-999-648 1 gal.
1 options
Catalog No. 19-999-648 Supplier STERIS™ Supplier No. 1J8108
Add to Cart
Add to Cart
Phosphoric acid-based de-scaler effectively removes scale deposits. Free-rinsing surgical instrument S-Klenz Scale Remover leaves no residue behind in automated washer/disinfectors. Easily measured and dispensed liquid formula.
S-Klenz Scale Remover
S-Klenz Scale Remover is a phosphoric acid-based film and scale remover. This instrument rust inhibitor is also used for de-scaling equipment.
How S-Klenz Scale Remover Works
Effective at removing lime scale, urine scale, and corrosion from stainless steel, glass, and all plastics. The wetting and penetrating agents of the formula remove film and scale, but is not recommended for use on aluminum surfaces, galvanized metals, and mild steel.
Why S-Klenz Scale Remover?
  • Productivity - Phosphoric acid-based de-scaler effectively removes scale deposits and rust.
  • Convenience - Easily measured and dispensed liquid formula.
  • Safety - Free-rinsing property leaves no residue behind on surgical instruments.
Specifications
Product Type Citric Acid-based Descaling Agent
Volume (English) 1 gal.
Volume (Metric) 3.78 L