Learn More
Medchemexpress LLC (S)-Lathosterol-d4 | 1182005-25-9 | 99.7% | 390.68 | 1 MG
SDP
Catalog No. 5000437483
Encompass_Preferred
(S)-Lathosterol-d4 is the deuterium labeled (S)-Lathosterol. Lathosterol is a cholesterol-like molecule, and serum Lathosterol concentration indicates whole-body cholesterol synthesis.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-37483 Supplier Medchemexpress LLC Supplier No. HY113486S1MG
(S)-Lathosterol-d4 is the deuterium labeled (S)-Lathosterol. Lathosterol is a cholesterol-like molecule, and serum Lathosterol concentration indicates whole-body cholesterol synthesis.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.