Learn More
Medchemexpress LLC (S)-Lathosterol-d4 | 1182005-25-9 | 99.7% | 390.68 | 500 UG
SDP
Catalog No. 5000436768
Encompass_Preferred
(S)-Lathosterol-d4 is a deuterium-labeled derivative of (S)-Lathosterol. Lathosterol itself is a molecule similar to cholesterol. The concentration of serum lathosterol is used as an indicator for the synthesis of cholesterol in the whole body. This compound is intended for research use only and is not sold to patients.
  • Deuterium labeled (S)-Lathosterol.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements can be incorporated into drug molecules for use as tracers in drug development.
  • Deuteration can potentially influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-36768 Supplier Medchemexpress LLC Supplier No. HY113486S500UG
(S)-Lathosterol-d4 is a deuterium-labeled derivative of (S)-Lathosterol. Lathosterol itself is a molecule similar to cholesterol. The concentration of serum lathosterol is used as an indicator for the synthesis of cholesterol in the whole body. This compound is intended for research use only and is not sold to patients.
  • Deuterium labeled (S)-Lathosterol.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements can be incorporated into drug molecules for use as tracers in drug development.
  • Deuteration can potentially influence the pharmacokinetic and metabolic profiles of drugs.