Learn More
Medchemexpress LLC (S)-Leucic acid | 13748-90-8 | 94.9% | 132.16 | 5 MG
SDP
Catalog No. 5000404133
Encompass_Preferred
(S)-Leucic acid (Standard) is the analytical standard of (S)-Leucic acid and is intended for research and analytical applications. It is a metabolite of Leucine and exhibits anabolic effects, particularly in promoting muscle protein synthesis. Additionally, (S)-Leucic acid can be utilized in the diagnosis of pregnancy loss. This compound is classified as an analytical standard and is commonly employed in qualitative, quantitative, and methodological research experiments using techniques such as HPLC, GC, and MS.
  • Analytical standard for (S)-Leucic acid
  • Intended for research and analytical applications
  • Metabolite of Leucine
  • Exhibits anabolic effects, promoting muscle protein synthesis
  • Can be utilized in the diagnosis of pregnancy loss
  • Commonly employed in qualitative, quantitative, and methodological research experiments
  • Used in techniques such as HPLC, GC, and MS
Catalog No. 50-004-04133 Supplier Medchemexpress LLC Supplier No. HY30215R5MG
(S)-Leucic acid (Standard) is the analytical standard of (S)-Leucic acid and is intended for research and analytical applications. It is a metabolite of Leucine and exhibits anabolic effects, particularly in promoting muscle protein synthesis. Additionally, (S)-Leucic acid can be utilized in the diagnosis of pregnancy loss. This compound is classified as an analytical standard and is commonly employed in qualitative, quantitative, and methodological research experiments using techniques such as HPLC, GC, and MS.
  • Analytical standard for (S)-Leucic acid
  • Intended for research and analytical applications
  • Metabolite of Leucine
  • Exhibits anabolic effects, promoting muscle protein synthesis
  • Can be utilized in the diagnosis of pregnancy loss
  • Commonly employed in qualitative, quantitative, and methodological research experiments
  • Used in techniques such as HPLC, GC, and MS