Learn More
Medchemexpress LLC (S)-Leucic acid | 13748-90-8 | 99.2% | 132.16 | 500 MG
SDP
Catalog No. 5000405055
Encompass_Preferred
(S)-Leucic acid is a metabolite of Leucine. It exhibits anabolic effects, specifically promoting muscle protein synthesis. Additionally, (S)-Leucic acid can be utilized in the diagnosis of pregnancy loss.
  • Metabolite of Leucine
  • Promotes muscle protein synthesis
  • Can be used in the diagnosis of pregnancy loss
  • Appears as a solid
  • Color is white to off-white
  • Shipped at room temperature in continental US
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-004-05055 Supplier Medchemexpress LLC Supplier No. HY30215500MG
(S)-Leucic acid is a metabolite of Leucine. It exhibits anabolic effects, specifically promoting muscle protein synthesis. Additionally, (S)-Leucic acid can be utilized in the diagnosis of pregnancy loss.
  • Metabolite of Leucine
  • Promotes muscle protein synthesis
  • Can be used in the diagnosis of pregnancy loss
  • Appears as a solid
  • Color is white to off-white
  • Shipped at room temperature in continental US
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month