Learn More
Medchemexpress LLC (S)-leucic acid | 13748-90-8 | MFCD00064214 | 94.9% | 132.16 g/mol | C6H12O3 | 10 MG
SDP
Catalog No. 5000297619
Encompass_Preferred
(S)-Leucic acid (Standard) is an analytical reference material of (S)-leucic acid (α-hydroxyisocaproic acid) supplied as a purified solid for research and analytical laboratory use. It is intended for assay calibration, method development, and reference testing in biochemical and analytical workflows.
  • Purified analytical standard with documented assay for reliable quantitation.
  • Suitable for HPLC and mass spectrometry method development and validation.
  • Available in small vial quantities to support analytical workflows.
  • Solid form with standard storage and handling characteristics for stability.
  • Certificate of analysis provided for batch-specific quality verification.
Catalog No. 50-002-97619 Supplier Medchemexpress LLC Supplier No. HY30215R10MG
(S)-Leucic acid (Standard) is an analytical reference material of (S)-leucic acid (α-hydroxyisocaproic acid) supplied as a purified solid for research and analytical laboratory use. It is intended for assay calibration, method development, and reference testing in biochemical and analytical workflows.
  • Purified analytical standard with documented assay for reliable quantitation.
  • Suitable for HPLC and mass spectrometry method development and validation.
  • Available in small vial quantities to support analytical workflows.
  • Solid form with standard storage and handling characteristics for stability.
  • Certificate of analysis provided for batch-specific quality verification.