Learn More
Medchemexpress LLC (S)-Leucic acid (Standard) | 13748-90-8 | 94.9% | 132.16 | 25 MG
SDP
Catalog No. 5000402731
Encompass_Preferred
(S)-Leucic acid (Standard) is the analytical standard of (S)-Leucic acid (HY-30215). This product is intended for research and analytical applications. (S)-Leucic acid is a metabolite of Leucine and has anabolic effects, particularly promoting muscle protein synthesis. It can also be used in the diagnosis of pregnancy loss.
  • The compound is an analytical standard, which is the reference standard supplied assay.
  • It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-02731 Supplier Medchemexpress LLC Supplier No. 25MGHY30215R
(S)-Leucic acid (Standard) is the analytical standard of (S)-Leucic acid (HY-30215). This product is intended for research and analytical applications. (S)-Leucic acid is a metabolite of Leucine and has anabolic effects, particularly promoting muscle protein synthesis. It can also be used in the diagnosis of pregnancy loss.
  • The compound is an analytical standard, which is the reference standard supplied assay.
  • It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.