Learn More
Medchemexpress LLC (S)-Leucic acid (standard) | 13748-90-8 | MFCD00064214 | 94.9% | 132.16 | 100 MG
SDP
Catalog No. 5000404371
Encompass_Preferred
(S)-Leucic acid (Standard) is an analytical standard used for research and analytical applications. As a metabolite of Leucine, it promotes muscle protein synthesis and can assist in the diagnosis of pregnancy loss. It is commonly utilized in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS.
  • Analytical standard
  • Intended for research and analytical applications
  • Metabolite of leucine
  • Promotes muscle protein synthesis
  • Can be used in the diagnosis of pregnancy loss
  • Suitable for HPLC, GC, and MS research experiments
Catalog No. 50-004-04371 Supplier Medchemexpress LLC Supplier No. HY30215R100MG
(S)-Leucic acid (Standard) is an analytical standard used for research and analytical applications. As a metabolite of Leucine, it promotes muscle protein synthesis and can assist in the diagnosis of pregnancy loss. It is commonly utilized in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS.
  • Analytical standard
  • Intended for research and analytical applications
  • Metabolite of leucine
  • Promotes muscle protein synthesis
  • Can be used in the diagnosis of pregnancy loss
  • Suitable for HPLC, GC, and MS research experiments