Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(-)-Limonene

Catalog No. 21836750G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-836-750G 50 g
21-836-7250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 21-836-750G Supplier Sigma Aldrich Supplier No. 21836750G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(-)-Limonene, a monoterpenoid compound, is mostly used in perfumery and in flavoring. On irradiation, it undergoes radiolysis, which leads to a reduction in the optical purity. (S)-(-)-Limonene also shows potent antimicrobial activity.

Specifications

CAS 5989-54-8
Density 0.844 g/mL at 25°C
Boiling Point 175°C to 177°C
Molecular Formula C10H16
Linear Formula C10H16
Refractive Index n20/D 1.471
MDL Number MFCD00001558
Quantity 50 g
Synonym (-)-p-Mentha-1,8-diene; (-)-Carvene; (S)-4-Isopropenyl-1-methyl cyclohexene
Molecular Weight (g/mol) 136.23 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.