Learn More
Cayman Chemical (S)-Lansoprazole, Purity: ≥98%, CAS: 138530-95-7, 10 mg
SDP
Catalog No. 502986446
Encompass_Preferred
A proton pump inhibitor and pure enantiomer of lansoprazole; inhibits H+/K+-stimulated ATPase pumps in parietal cells (IC50 = 5.2 µM); inhibits acid formation in isolated canine parietal cells (IC50 = 82 µM)
Catalog No. 50-298-6446 Supplier Cayman Chemical Supplier No. 2196710 MG
A proton pump inhibitor and pure enantiomer of lansoprazole; inhibits H+/K+-stimulated ATPase pumps in parietal cells (IC50 = 5.2 µM); inhibits acid formation in isolated canine parietal cells (IC50 = 82 µM)