Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC S-malic acid-13C4 | 150992-96-4 | 98.0% | 138.06 g·mol⁻1 | 13C4H6O5 | 10mg
SDP

Catalog No. 5000267681
Encompass_Preferred

(S)-Malic acid-13C4 is the 13C labeled S-Malic acid (HY-Y1069)[1]

Catalog No. 50-002-67681 Supplier Medchemexpress LLC Supplier No. HYY1069S310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-Malic acid-13C4 is the 13C labeled S-Malic acid (HY-Y1069)[1]

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.