Learn More
Medchemexpress LLC HY-N2176 5mg Medchemexpress, S-(+)-Marmesin CAS:13849-08-6 Purity:>98%
SDP
Catalog No. 502082873
Encompass_Preferred
Medchemexpress, HY-N2176 5mg S-(+)-Marmesin CAS:13849-08-6 S-(+)-Marmesin is a natural coumarin, exhibiting COX-2/5-LOX dual inhibitory activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2873 Supplier Medchemexpress LLC Supplier No. HYN21765MG
Medchemexpress, HY-N2176 5mg S-(+)-Marmesin CAS:13849-08-6 S-(+)-Marmesin is a natural coumarin, exhibiting COX-2/5-LOX dual inhibitory activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.