Learn More
Cayman Chemical S-MephenytoIn 10mg
SDP
Catalog No. 502955783
Encompass_Preferred
A substrate of the CYP450 isoform CYP2C19, also known as mephenytoin 4-hydroxylase; can be used to screen for mutations in CYP2C19 by assaying its metabolites in urine
Catalog No. 50-295-5783 Supplier Cayman Chemical Supplier No. 1191310 MG
A substrate of the CYP450 isoform CYP2C19, also known as mephenytoin 4-hydroxylase; can be used to screen for mutations in CYP2C19 by assaying its metabolites in urine