Learn More
Enzo Life Sciences (S)-Mephenytoin, (25mg), CAS Number: 70989-04-7
SDP
Supplier:  Enzo Life Sciences BMLJ1200025
Encompass_Preferred
Cytochrome P450 substrate. Substrate for determination of cytochrome P-450 polymorphism. Anticonvulsant. Formula: C12H14N2O2. MW: 218.3. CAS Number: 70989-04-7. Purity: ≥98% (TLC). Appearance: White solid. Solubility: Soluble in DMSO or ethanol. Long Term Storage: Ambient
Catalog No. NC9032495