Learn More
TARGETMOL CHEMICALS INC S-Mephenytoin 5MG
SDP
Catalog No. 502875997
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. (S)-Mephenytoin ((+)-Mephenytoin) is an anticonvulsant that serves as a substrate of the cytochrome P450 (CYP) isoform CYP2C19 and is used to analyze cytochrome P450 metabolism.
Catalog No. 50-287-5997 Supplier TARGETMOL CHEMICALS INC Supplier No. T215295MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. (S)-Mephenytoin ((+)-Mephenytoin) is an anticonvulsant that serves as a substrate of the cytochrome P450 (CYP) isoform CYP2C19 and is used to analyze cytochrome P450 metabolism.