Learn More
Medchemexpress LLC (S)-Mephenytoin | 70989-04-7 | MFCD00270025 | 100.0% | 218.25 g·mol-1 | C12H14N2O2 | 10 MG
SDP
Catalog No. 5000299011
Encompass_Preferred
(S)-Mephenytoin is the S-enantiomer of the anticonvulsant mephenytoin used as a probe substrate for cytochrome P450 enzymes, particularly CYP2C19. It is intended for research use in metabolic assays, enzyme kinetics, and analytical studies, supplied as a white solid with accompanying purity data.
  • High enantiomeric purity (≈99.9% ee)
  • Suitable as a CYP2C19 substrate for metabolic assays
  • Available in small mass quantities for analytical and reference use
  • Provided as a stable white solid with supporting datasheet and purity information
Catalog No. 50-002-99011 Supplier Medchemexpress LLC Supplier No. HYB1184A10MG
(S)-Mephenytoin is the S-enantiomer of the anticonvulsant mephenytoin used as a probe substrate for cytochrome P450 enzymes, particularly CYP2C19. It is intended for research use in metabolic assays, enzyme kinetics, and analytical studies, supplied as a white solid with accompanying purity data.
  • High enantiomeric purity (≈99.9% ee)
  • Suitable as a CYP2C19 substrate for metabolic assays
  • Available in small mass quantities for analytical and reference use
  • Provided as a stable white solid with supporting datasheet and purity information