Learn More
Medchemexpress LLC (S)-mephenytoin | 70989-04-7 | MFCD00270025 | 99.9% | 218.25 g/mol | C12H14N2O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB1184AR10MG
Encompass_Preferred
(S)-Mephenytoin is the S-enantiomer of mephenytoin supplied as an analytical reference standard for research and analytical applications. It is commonly used as a substrate for cytochrome P450 (CYP) 2C19 assays, for method development, and for quality control. Typical specifications include high purity (≈99.9%) and a molecular weight of 218.25 g/mol.
  • Analytical standard suitable for reference and quality control.
  • High purity for accurate analytical measurements.
  • Used as a substrate in CYP2C19 activity assays.
  • Available in small pack sizes for analytical workflows.
  • White to off-white solid, suitable for laboratory handling.
Catalog No. 50-002-97746