Learn More
Medchemexpress LLC (S)-Mephenytoin ((+)-Mephenytoin) | 70989-04-7 | 99.9% | 100 MG
SDP
Catalog No. 5000411904
Encompass_Preferred
(S)-Mephenytoin is an anticonvulsive agent and a substrate of the cytochrome P450 (CYP) isoform CYP2C19. This compound is primarily utilized for the analysis of cytochrome P450 metabolism in research applications.
  • Anticonvulsive properties
  • Acts as a substrate for CYP2C19 isoform
  • Supports analysis of cytochrome P450 metabolism
  • High purity for reliable research
  • Suitable for in vitro and in vivo studies
Catalog No. 50-004-11904 Supplier Medchemexpress LLC Supplier No. HYB1184A100MG
(S)-Mephenytoin is an anticonvulsive agent and a substrate of the cytochrome P450 (CYP) isoform CYP2C19. This compound is primarily utilized for the analysis of cytochrome P450 metabolism in research applications.
  • Anticonvulsive properties
  • Acts as a substrate for CYP2C19 isoform
  • Supports analysis of cytochrome P450 metabolism
  • High purity for reliable research
  • Suitable for in vitro and in vivo studies