Learn More
Medchemexpress LLC (S)-Mephenytoin ((+)-Mephenytoin) | 70989-04-7 | 99.9% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB1184A5MG
Encompass_Preferred
(S)-Mephenytoin is an anticonvulsive agent. It is a substrate of the cytochrome P450 (CYP) isoform CYP2C19 and can be used for the analysis of cytochrome P450 metabolism.
  • Anticonvulsive agent
  • Substrate of the cytochrome P450 (CYP) isoform CYP2C19
  • Can be used for the analysis of cytochrome P450 metabolism
  • For research use only
Catalog No. 50-004-13450