Learn More
Medchemexpress LLC (S)-Mephenytoin (Standard) | 70989-04-7 | 99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB1184AR100MG
Encompass_Preferred
(S)-Mephenytoin (Standard) is an analytical standard intended for research and analytical applications. It is an anticonvulsive agent and a substrate of the cytochrome P450 (CYP) isoform CYP2C19. This compound is a grade of analytical standard used in qualitative, quantitative, and methodological research experiments.
  • Used for the analysis of cytochrome P450 metabolism.
  • Suitable for applications in HPLC, GC, and MS.
Catalog No. 50-004-12409