Learn More
Medchemexpress LLC HY-B1161A 100mg , S-Methoprene CAS:65733-16-6 Purity:>98%
SDP
Catalog No. 502261929
Encompass_Preferred
HY-B1161A 100mg S-Methoprene CAS: 65733-16-6 (S)-Methoprene is a juvenile hormone analogue that prohibits the ability of the insect to change from pupae to adult and is used as an insecticide. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-1929 Supplier Medchemexpress LLC Supplier No. HYB1161A100MG
HY-B1161A 100mg S-Methoprene CAS: 65733-16-6 (S)-Methoprene is a juvenile hormone analogue that prohibits the ability of the insect to change from pupae to adult and is used as an insecticide. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.