Learn More
Medchemexpress LLC (S)-methyl 2-(6-hydroxy-2,3-dihydrobenzofuran-3-yl)acetate | 1000414-38-9 | 98.8% | C11H12O4 | 250 MG
SDP
Catalog No. 5000358689
Encompass_Preferred
(S)-methyl 2-(6-hydroxy-2,3-dihydrobenzofuran-3-yl)acetate is a chemical compound provided by MedChemExpress, catalog number HY-76785. Primarily used for laboratory research and the manufacture of substances, it is supplied as a solid with a purity of 98.8%.
  • For laboratory chemicals and substance manufacturing.
  • Supplied as a solid.
  • Purity of 98.8%.
  • Powder storage: -20°C for 3 years, 4°C for 2 years.
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month.
  • Room temperature shipping for less than 2 weeks.
  • Harmful if swallowed, causes skin and serious eye irritation.
Catalog No. 50-003-58689 Supplier Medchemexpress LLC Supplier No. 250MGHY76785
(S)-methyl 2-(6-hydroxy-2,3-dihydrobenzofuran-3-yl)acetate is a chemical compound provided by MedChemExpress, catalog number HY-76785. Primarily used for laboratory research and the manufacture of substances, it is supplied as a solid with a purity of 98.8%.
  • For laboratory chemicals and substance manufacturing.
  • Supplied as a solid.
  • Purity of 98.8%.
  • Powder storage: -20°C for 3 years, 4°C for 2 years.
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month.
  • Room temperature shipping for less than 2 weeks.
  • Harmful if swallowed, causes skin and serious eye irritation.