Learn More
Medchemexpress LLC S-methyl-KE-298 | 143584-75-2 | ≥98.0% | 252.33 g/mol | C13H16O3S | 25 MG
SDP
Catalog No. 5000248674
Encompass_Preferred
S-methyl-KE-298 is the S-methylated active metabolite of KE-298, reported to inhibit matrix metalloproteinase-1 (MMP-1) production in rheumatoid arthritis synovial cells. It is supplied as a purified research chemical (solid) for laboratory assays and analytical work.
  • Reported inhibitor of matrix metalloproteinase-1 (MMP-1) production in RA synovial cells.
  • High purity (≥98.0%) suitable for analytical and biological research.
  • Molecular weight 252.33 g/mol and chemical formula C13H16O3S.
  • Available in small research pack sizes such as 25 mg and recommended refrigerated storage.
  • Product page includes a downloadable datasheet with characterization data.
Catalog No. 50-002-48674 Supplier Medchemexpress LLC Supplier No. HY10167125MG
S-methyl-KE-298 is the S-methylated active metabolite of KE-298, reported to inhibit matrix metalloproteinase-1 (MMP-1) production in rheumatoid arthritis synovial cells. It is supplied as a purified research chemical (solid) for laboratory assays and analytical work.
  • Reported inhibitor of matrix metalloproteinase-1 (MMP-1) production in RA synovial cells.
  • High purity (≥98.0%) suitable for analytical and biological research.
  • Molecular weight 252.33 g/mol and chemical formula C13H16O3S.
  • Available in small research pack sizes such as 25 mg and recommended refrigerated storage.
  • Product page includes a downloadable datasheet with characterization data.