Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC S-methyl-KE-298 | 143584-75-2 | MFCD31382145 | >=98.0% | 252.33 g/mol | C13H16O3S | 5 MG
SDP

Catalog No. 5000247039
Encompass_Preferred

S-methyl-KE-298 is the S-methylated active metabolite of KE-298 reported to inhibit matrix metalloproteinase-1 (MMP-1) production from rheumatoid arthritis (RA) synovial cells. It is supplied as a research-grade small molecule for biochemical and cellular studies.

  • Active metabolite with reported MMP-1 inhibitory activity
  • Purity ≥98.0% (HPLC)
  • Chemical formula C13H16O3S; molecular weight 252.33 g/mol
  • Available in milligram quantities (e.g., 5 mg)
  • Soluble in DMSO for assay preparation
  • Intended for research use in biochemical and cellular assays

Catalog No. 50-002-47039 Supplier Medchemexpress LLC Supplier No. HY1016715MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S-methyl-KE-298 is the S-methylated active metabolite of KE-298 reported to inhibit matrix metalloproteinase-1 (MMP-1) production from rheumatoid arthritis (RA) synovial cells. It is supplied as a research-grade small molecule for biochemical and cellular studies.

  • Active metabolite with reported MMP-1 inhibitory activity
  • Purity ≥98.0% (HPLC)
  • Chemical formula C13H16O3S; molecular weight 252.33 g/mol
  • Available in milligram quantities (e.g., 5 mg)
  • Soluble in DMSO for assay preparation
  • Intended for research use in biochemical and cellular assays

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.