Learn More
Medchemexpress LLC S-methyl-KE-298 | 143584-75-2 | MFCD43584752 | 98.0% | 252.33 g/mol | C13H16O3S | 50 MG
SDP
Catalog No. 5000249103
Encompass_Preferred
S-methyl-KE-298 is the S-methylated active metabolite of KE-298 used in research on matrix metalloproteinase (MMP) regulation. It is applied in in vitro studies to evaluate metabolism, protein binding, and effects on MMP-1 and MT1-MMP expression relevant to rheumatoid arthritis synovial cells.
  • Active metabolite useful for MMP biology studies.
  • Characterized by analytical data including molecular formula C13H16O3S and molecular weight 252.33 g/mol.
  • High purity suitable for analytical and in vitro applications.
  • Reported high plasma protein binding in preclinical species and human plasma.
  • Available in small milligram quantities for research workflows.
Catalog No. 50-002-49103 Supplier Medchemexpress LLC Supplier No. HY10167150MG
Add to Cart
Add to Cart
S-methyl-KE-298 is the S-methylated active metabolite of KE-298 used in research on matrix metalloproteinase (MMP) regulation. It is applied in in vitro studies to evaluate metabolism, protein binding, and effects on MMP-1 and MT1-MMP expression relevant to rheumatoid arthritis synovial cells.
  • Active metabolite useful for MMP biology studies.
  • Characterized by analytical data including molecular formula C13H16O3S and molecular weight 252.33 g/mol.
  • High purity suitable for analytical and in vitro applications.
  • Reported high plasma protein binding in preclinical species and human plasma.
  • Available in small milligram quantities for research workflows.