Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich S-Methyl methanethiosulfonate
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Interaction of s-methyl methanethiosulfonate (mmts) with dipalmitoylphosphatidylcholine (dppc) bilayers has been investigated by ftir and surface-enhanced raman spectroscopy.s-methyl methanethiosulfonate was used as sulfenylating agent for α-keto sulfoxides, methylene compounds, half-esters of malonic acids and aryl grignard reagents
- It was also used as a reagent to trap the natural thiol-disulfide state of the proteins
Specifications
Specifications
| CAS | 2949-92-0 |
| Density | 1.337 g/mL (at 25°C (literature)) |
| Boiling Point | 69°C to 71°C (0.4 mmHg) |
| Molecular Formula | C2H6O2S2 |
| Refractive Index | n20/D 1.513 (literature) |
| Linear Formula | CH3SO2SCH3 |
| MDL Number | MFCD00007565 |
| Quantity | 10 g |
| Synonym | S-Methyl thiomethanesulfonate; MMTS |
| Molecular Weight (g/mol) | 126.2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?