Alfa Aesar™ S-Methyl thiobutyrate, 98%

Manufacturer: Alfa Aesar™ B2214406

Quantity 5g

 View more versions of this product

Catalog No. AAB2214406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2432-51-1
    Molecular Formula C5H10OS
    Formula Weight 118.2
    MDL Number MFCD00009872
    UN Number UN1993
    Beilstein 1848987
    Synonym S-Methyl thiobutanoate
    Chemical Name or Material S-Methyl thiobutyrate

    Specifications

    Formula Weight 118.2
    TSCA TSCA
    Molecular Formula C5H10OS
    UN Number UN1993
    Synonym S-Methyl thiobutanoate
    Flash Point 34°C (93°F)
    MDL Number MFCD00009872
    CAS 2432-51-1
    Beilstein 1848987
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 142° to 143°C
    Quantity 5g
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Chemical Name or Material S-Methyl thiobutyrate
    Assay 98%

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit