Learn More
Medchemexpress LLC S-Methyl thiomethanesulfonate (MMTS) | 2949-92-0 | 126.20 | 100 G
SDP
Catalog No. 5000445413
Encompass_Preferred
S-Methyl thiomethanesulfonate (MMTS) is a sulfur-containing volatile organic compound produced by plants and bacteria. It is an effective anti-oomycete agent and finds applications in infection research, as a metabolic enzyme/protease, and in anti-infection studies.
  • Sulfur-containing volatile organic compound
  • Produced by plants and bacteria
  • Effective anti-oomycete agent
  • Applications in infection research
  • Related to metabolic enzyme/protease studies
  • Used in anti-infection applications
  • Functions as an endogenous metabolite
  • Involved in bacterial studies
Catalog No. 50-004-45413 Supplier Medchemexpress LLC Supplier No. HYW004461100G
S-Methyl thiomethanesulfonate (MMTS) is a sulfur-containing volatile organic compound produced by plants and bacteria. It is an effective anti-oomycete agent and finds applications in infection research, as a metabolic enzyme/protease, and in anti-infection studies.
  • Sulfur-containing volatile organic compound
  • Produced by plants and bacteria
  • Effective anti-oomycete agent
  • Applications in infection research
  • Related to metabolic enzyme/protease studies
  • Used in anti-infection applications
  • Functions as an endogenous metabolite
  • Involved in bacterial studies