Learn More
(S)-(-)-α-Methylbenzyl isocyanate, ≥99.0% (sum of enantiomers, GC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| CAS | 14649-03-7 |
| Density | 1.045 g/mL (at 20°C) |
| Boiling Point | 55°C to 56°C/2.5 mmHg |
| Flash Point | 65°C - Closed Cup |
| Linear Formula | C6H5 CH(CH3)NCO |
| Quantity | 5 mL |
| Formula Weight | 147.17 |
| Percent Purity | ≥99.0% (sum of enantiomers, GC); ≥99.0% |
| Grade | For chiral derivatization |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.