Learn More
Medchemexpress LLC HY-W009177 5mg Medchemexpress, S-Methylglutathione CAS:2922-56-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW0091775MG
Encompass_Preferred
Medchemexpress, HY-W009177 5mg S-Methylglutathione CAS:2922-56-7 S-Methylglutathione is an S-substitued glutathione and a stronger nucleophile than GSH. S-Methylglutathione has inhibitory effect on glyoxalase 1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3266