Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich S-Methylisothiourea hemisulfate salt

Catalog No. M84445500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M84445500G 500 g
M84445100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M84445500G Supplier Sigma Aldrich Supplier No. M84445500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

S-Methylisothiourea hemisulfate salt

Specifications

CAS 867-44-7
Melting Point 240°C to 241°C (dec.) (lit.)
Molecular Formula C2H6N2S -+ 0.5 H2SO4
Linear Formula HN=C(NH2)SCH3 -+ 1/2H2SO4
Quantity 500 g
MDL Number MFCD00013055
Synonym 2-Methyl-2-thiopseudourea hemisulfate salt; S-Methyl-ITU; S-Methylthiuronium sulfate; SMT
Molecular Weight (g/mol) 139.19
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.