Learn More
Medchemexpress LLC HY-79457 25mg Medchemexpress, S-Methylisothiourea (sulfate) CAS:867-44-7 Purity:>98%
SDP
Catalog No. 502082295
Encompass_Preferred
Medchemexpress, HY-79457 25mg S-Methylisothiourea (sulfate) CAS:867-44-7 S-Methylisothiourea sulfate is a potent, selective and competitive inhibitor of inducible nitric oxide synthase (iNOS). S-Methylisothiourea sulfate exerts beneficial effects in rodent models of septic shock. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2295 Supplier Medchemexpress LLC Supplier No. HY7945725MG
Medchemexpress, HY-79457 25mg S-Methylisothiourea (sulfate) CAS:867-44-7 S-Methylisothiourea sulfate is a potent, selective and competitive inhibitor of inducible nitric oxide synthase (iNOS). S-Methylisothiourea sulfate exerts beneficial effects in rodent models of septic shock. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.