Learn More
Apexbio Technology LLC (S)-Methylisothiourea sulfate 867-44-7 50mg
SDP
Catalog No. 5000088990
Encompass_Preferred
S-Methylisothiourea sulfate (CAS 867-44-7) is a small-molecule inhibitor targeting inducible nitric oxide synthase (iNOS) It is designed to selectively and competitively inhibit iNOS thereby modulating nitric oxide (NO) synthesis under inflammatory conditions S-Methylisothiourea sulfate exerts its biological activity primarily through direct inhibition of iNOS The compound demonstrates an inhibitory activity profile with reported IC50 values for iNOS inhibition in the range of approximately 7 10 M Based on these pharmacological properties S-Methylisothiourea sulfate holds research potential in investigating inducible nitric oxide signaling pathways and exploring therapeutic approaches for inflammation-driven pathologies including septic shock and autoimmune disorders
Catalog No. 50-000-88990 Supplier Apexbio Technology LLC Supplier No. B644750
S-Methylisothiourea sulfate (CAS 867-44-7) is a small-molecule inhibitor targeting inducible nitric oxide synthase (iNOS) It is designed to selectively and competitively inhibit iNOS thereby modulating nitric oxide (NO) synthesis under inflammatory conditions S-Methylisothiourea sulfate exerts its biological activity primarily through direct inhibition of iNOS The compound demonstrates an inhibitory activity profile with reported IC50 values for iNOS inhibition in the range of approximately 7 10 M Based on these pharmacological properties S-Methylisothiourea sulfate holds research potential in investigating inducible nitric oxide signaling pathways and exploring therapeutic approaches for inflammation-driven pathologies including septic shock and autoimmune disorders