Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC S-mirtazapine | 61337-87-9 | MFCD00006761 | 99.8% | 265.35 g/mol | C17H19N3 | 10 MG
SDP

Catalog No. 5000298086
Encompass_Preferred

(S)-Mirtazapine is the S(+)-enantiomer of mirtazapine, provided as a solid research chemical for laboratory and analytical use. For research use only; not for human or clinical use.

  • High purity: 99.76% (enantiomeric excess 98.35%).
  • Chemical formula: C17H19N3; molecular weight: 265.35 g/mol.
  • Appearance: off-white to light yellow powder.
  • Solubility: ≥100 mg/mL in DMSO; use newly opened DMSO due to hygroscopicity.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Pack sizes suitable for small-scale research, including 10 mg quantities.

Catalog No. 50-002-98086 Supplier Medchemexpress LLC Supplier No. HYB0352A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-Mirtazapine is the S(+)-enantiomer of mirtazapine, provided as a solid research chemical for laboratory and analytical use. For research use only; not for human or clinical use.

  • High purity: 99.76% (enantiomeric excess 98.35%).
  • Chemical formula: C17H19N3; molecular weight: 265.35 g/mol.
  • Appearance: off-white to light yellow powder.
  • Solubility: ≥100 mg/mL in DMSO; use newly opened DMSO due to hygroscopicity.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Pack sizes suitable for small-scale research, including 10 mg quantities.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.