Learn More
Medchemexpress LLC S-Mirtazapine-d3 | 61337-87-9 | 99.0% | 1 MG
SDP
Catalog No. 5000405314
Encompass_Preferred
(S)-Mirtazapine-d3 is a deuterium labeled (S)-Mirtazapine, which is an S(+)-enantiomer of Mirtazapine. It exhibits pronociceptive properties in an animal model of acute thermal nociception and acts as a stereoselective 5-HT2 receptor antagonist. This compound is metabolized by CYP2D6 and CYP1A2.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stereoselective 5-HT2 receptor antagonist.
  • Metabolized by CYP2D6 and CYP1A2.
Catalog No. 50-004-05314 Supplier Medchemexpress LLC Supplier No. HYB0352AS1MG
(S)-Mirtazapine-d3 is a deuterium labeled (S)-Mirtazapine, which is an S(+)-enantiomer of Mirtazapine. It exhibits pronociceptive properties in an animal model of acute thermal nociception and acts as a stereoselective 5-HT2 receptor antagonist. This compound is metabolized by CYP2D6 and CYP1A2.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stereoselective 5-HT2 receptor antagonist.
  • Metabolized by CYP2D6 and CYP1A2.