Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC S-mirtazapine-d3 | 61337-87-9 | 99.0% | 268.37 | C17H16D3N3 | 10 MG
SDP

Catalog No. 5000299040
Encompass_Preferred

Deuterated S-mirtazapine is the S-enantiomer of mirtazapine labeled with three deuterium atoms, supplied as a research-grade reference standard for analytical and pharmacology studies.

  • Isotopically labeled internal standard for LC-MS/MS quantitation.
  • High purity for reliable analytical results.
  • Molecular weight 268.37 and formula C17H16D3N3.
  • Supplied as a 10 mg vial for method development and calibration.

Catalog No. 50-002-99040 Supplier Medchemexpress LLC Supplier No. HYB0352AS10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deuterated S-mirtazapine is the S-enantiomer of mirtazapine labeled with three deuterium atoms, supplied as a research-grade reference standard for analytical and pharmacology studies.

  • Isotopically labeled internal standard for LC-MS/MS quantitation.
  • High purity for reliable analytical results.
  • Molecular weight 268.37 and formula C17H16D3N3.
  • Supplied as a 10 mg vial for method development and calibration.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.