Learn More
Medchemexpress LLC (S)-Mirtazapine-d3 | 61337-87-9 | 99.0% | 5 MG
SDP
Catalog No. 5000406110
Encompass_Preferred
(S)-Mirtazapine-d3, also known as (S)-Org3770 d3 or (S)-6-Azamianserin d3, is a deuterium-labeled compound. It acts as an 5-HT2 receptor antagonist and has a purity of 99.0%. The molecular formula is C17H16D3N3 and its molecular weight is 268.37.
  • Deuterium-labeled compound
  • Acts as an 5-HT2 receptor antagonist
  • High purity of 99.0%
  • Identified by molecular formula C17H16D3N3
  • Molecular weight of 268.37
Catalog No. 50-004-06110 Supplier Medchemexpress LLC Supplier No. HYB0352AS5MG
(S)-Mirtazapine-d3, also known as (S)-Org3770 d3 or (S)-6-Azamianserin d3, is a deuterium-labeled compound. It acts as an 5-HT2 receptor antagonist and has a purity of 99.0%. The molecular formula is C17H16D3N3 and its molecular weight is 268.37.
  • Deuterium-labeled compound
  • Acts as an 5-HT2 receptor antagonist
  • High purity of 99.0%
  • Identified by molecular formula C17H16D3N3
  • Molecular weight of 268.37