Learn More
Medchemexpress LLC HY-100390 5mg Medchemexpress, (S)-ML753286 CAS:1699720-85-8 Purity:>98%
SDP
Catalog No. 501931806
Encompass_Preferred
Medchemexpress, HY-100390 5mg (S)-ML753286 CAS:1699720-85-8 (S)-ML753286 is a breast cancer resistance protein ( BCRP ) inhibitor with an IC 50 of 0.6 μM on BCRP efflux transporter. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1806 Supplier Medchemexpress LLC Supplier No. HY1003905MG
Medchemexpress, HY-100390 5mg (S)-ML753286 CAS:1699720-85-8 (S)-ML753286 is a breast cancer resistance protein ( BCRP ) inhibitor with an IC 50 of 0.6 μM on BCRP efflux transporter. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.