Learn More
Sarstedt Inc S-Monovette, 1.2mL, EDTA, Transparent, 50/Pack
Supplier:  Sarstedt Inc 06.1664.100
Encompass_Preferred
Haematology potassium EDTA, S-Monovette (needles, Multifly needles and adapters), The highly transparent plastic materials used in the manufacture of the S-Monovette ensure optimal visual assessment, 1.2mL
Catalog No. NC9345109