Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-(2-Benzoylphenyl)-1-benzylpyrrolidine-2-carboxamide 98.0+%, TCI America™
SDP

Catalog No. B55191G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B55191G 1 g
B55195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B55191G Supplier TCI America Supplier No. B55191G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 96293-17-3
Molecular Formula C25H24N2O2
Molecular Weight (g/mol) 384.479
MDL Number MFCD00145260
InChI Key IPSABLMEYFYEHS-QHCPKHFHSA-N
PubChem CID 980835
IUPAC Name (2S)-N-(2-benzoylphenyl)-1-benzylpyrrolidine-2-carboxamide
SMILES C1CC(N(C1)CC2=CC=CC=C2)C(=O)NC3=CC=CC=C3C(=O)C4=CC=CC=C4

Specifications

Melting Point 100°C
Color White-Yellow
Quantity 1 g
Formula Weight 384.48
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-N-(2-Benzoylphenyl)-1-benzylpyrrolidine-2-carboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.