Learn More
Medchemexpress LLC (S)-N-((3-Methyl-1-(methyl-d3)-1H-pyrazol-4-yl)sulfonyl)-6-(3-(3,3,3-trifluoro-2,2-dimethylpropoxy)-1H-pyrazol-1-yl)-2-(2,2,4-trimethylpyrrolidin-1-yl)nicotinamide | 2216712-66-0 | 99.23% | 600.67 | 500 UG
SDP
Catalog No. 5000438896
Encompass_Preferred
Elexacaftor-d3 (VX-445-d3) is the deuterium labeled form of Elexacaftor, designed as a tracer and internal standard for quantitative analysis using methods like NMR, GC-MS, or LC-MS. Deuterium substitution can impact the pharmacokinetic and metabolic profiles of drugs. Elexacaftor (VX-445) is a next-generation cystic fibrosis transmembrane conductance regulator (CFTR) corrector, which helps restore Phe508del CFTR protein function and has potential in treating cystic fibrosis. When combined with Tezacaftor and VX-770, it significantly enhances Phe508del CFTR protein processing, trafficking, and chloride transport.
  • Tracer and internal standard for quantitative analysis
  • Potential to influence pharmacokinetic and metabolic profiles of drugs
  • Restores Phe508del CFTR protein function
  • Potential to treat cystic fibrosis
  • Enhances CFTR protein processing, trafficking, and chloride transport
Catalog No. 50-004-38896 Supplier Medchemexpress LLC Supplier No. HY111772S1500UG
Elexacaftor-d3 (VX-445-d3) is the deuterium labeled form of Elexacaftor, designed as a tracer and internal standard for quantitative analysis using methods like NMR, GC-MS, or LC-MS. Deuterium substitution can impact the pharmacokinetic and metabolic profiles of drugs. Elexacaftor (VX-445) is a next-generation cystic fibrosis transmembrane conductance regulator (CFTR) corrector, which helps restore Phe508del CFTR protein function and has potential in treating cystic fibrosis. When combined with Tezacaftor and VX-770, it significantly enhances Phe508del CFTR protein processing, trafficking, and chloride transport.
  • Tracer and internal standard for quantitative analysis
  • Potential to influence pharmacokinetic and metabolic profiles of drugs
  • Restores Phe508del CFTR protein function
  • Potential to treat cystic fibrosis
  • Enhances CFTR protein processing, trafficking, and chloride transport