Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-N-(alpha-Methylbenzyl)phthalamic Acid 98.0+%, TCI America™
SDP

Catalog No. M08241G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M08241G Supplier TCI America Supplier No. M08241G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 21752-36-3
Molecular Formula C16H15NO3
Molecular Weight (g/mol) 269.3
MDL Number MFCD00013979
InChI Key VCFKXWGKKDZMPO-NSHDSACASA-N
Synonym s-2-1-phenylethyl carbamoyl benzoic acid, s---n-alpha-methylbenzyl phthalamic acid, s---n-1-phenylethyl phthalamic acid, 2-1s-1-phenylethyl carbamoyl benzoic acid, s-o-1-phenylethyl amino carbonyl benzoic acid, benzoic acid,2-1s-1-phenylethyl amino carbonyl, s---n-alpha-methylbenzyl phthalamicacid, s---n-, a-methylbenzyl phthalamic acid, s---n-a-methylbenzyl phthalamidic acid
PubChem CID 89037
IUPAC Name 2-[[(1S)-1-phenylethyl]carbamoyl]benzoic acid
SMILES CC(C1=CC=CC=C1)NC(=O)C2=CC=CC=C2C(=O)O

Specifications

Melting Point 132°C
Color White
Quantity 1 g
Formula Weight 269.30
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-N-(alpha-Methylbenzyl)phthalamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.