Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(-)-N-Benzyl-Alpha-Methylbenzylamine

Catalog No. 42747050ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-747-050ML 50 mL
42-747-010ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 42-747-050ML Supplier Sigma Aldrich Supplier No. 42747050ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(-)-N-Benzyl-+a-methylbenzylamine can be used:In one of the key synthetic steps for the preparation of a cardioprotective drug named CP-060S.To prepare (1S,2S,3S,5R)-tert-butyl 3-[benzyl((S)-1-phenylethyl)amino]-6,6-dimethylbicyclo[3.1.1]heptane-2-carboxylate, an intermediate used to synthesize pinane-based +a- and +a-amino acids.To prepare urea derivatives as potent antimicrobial agents.

Specifications

CAS 17480-69-2
Density 1.01 g/mL at 25°C
Boiling Point 171°C (15 mmHg, lit.)
Molecular Formula C15H17N
Refractive Index n20/D 1.563
Linear Formula C6H5CH(CH3)NHCH2C6H5
MDL Number MFCD00066325
Quantity 50 mL
Synonym (S)-(-)-N-(1-Phenylethyl)benzylamine; (S)-(-)-N-Benzyl-alpha-phenylethylamine
Molecular Weight (g/mol) 211.3 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.