Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-N-Boc-1,2,3,6-Tetrahydro-2-Pyridinecarboxylic Acid

Catalog No. 6702861G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-028-61G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 67-028-61G Supplier Sigma Aldrich Supplier No. 6702861G
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-N-Boc-1,2,3,6-Tetrahydro-2-Pyridinecarboxylic Acid

Specifications

CAS 7408-81-3
Melting Point 139°C to 143°C
Molecular Formula C13H27NO2
Linear Formula CH3NH(CH2)11COOH
MDL Number MFCD00042652
Quantity 5 g
Synonym (S)-N-Boc-1,2,3,6-tetrahydro-2-picolinic acid
Molecular Weight (g/mol) 229.36 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.