Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-Boc-2-hydroxymethylmorpholine, 98%

Catalog No. AAH6068003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6068003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6068003 Supplier Thermo Scientific Chemicals Supplier No. H6068003
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 135065-76-8 | C10H19NO4 | 217.265 g/mol

(S)-N-Boc-2-hydroxymethylmorpholine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-N-Boc-2-hydroxymethylmorpholine is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Store away from oxidizing agents, air, acids. Store in dark
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135065-76-8
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.265
MDL Number MFCD09260606
InChI Key FJYBLMJHXRWDAQ-QMMMGPOBSA-N
Synonym s-n-boc-2-hydroxymethylmorpholine, s-tert-butyl 2-hydroxymethyl morpholine-4-carboxylate, tert-butyl 2s-2-hydroxymethyl morpholine-4-carboxylate, s-4-boc-2-hydroxymethyl-morpholine, s-2-hydroxymethyl-morpholine-4-carboxylic acid tert-butyl ester, s-2-hydroxymethylmorpholine-4-carboxylic acid tert-butyl ester, s-n-boc-2-hydroxymethyl morpholine, 2s-2-hydroxymethyl-morpholine-4-carboxylic acid tert-butyl ester, pubchem18048, pubchem18049
PubChem CID 1512577
IUPAC Name tert-butyl (2S)-2-(hydroxymethyl)morpholine-4-carboxylate
SMILES CC(C)(C)OC(=O)N1CCOC(C1)CO

Specifications

Solubility Information Slightly soluble in water.
Quantity 1 g
Formula Weight 217.27
Sensitivity Light sensitive
Percent Purity 98%
Chemical Name or Material (S)-N-Boc-2-hydroxymethylmorpholine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.