Learn More
Medchemexpress LLC (S)-N-Boc-3-hydroxypyrrolidine | 101469-92-5 | 99.98% | C9H17NO3 | 5 G
SDP
Catalog No. 5000434096
Encompass_Preferred
(S)-N-Boc-3-hydroxypyrrolidine is a pharmaceutical intermediate. It can be used for the synthesis of other compounds and can be derivatized to yield 5-HT1A partial agonists.
  • Used for the synthesis of other compounds
  • Can be derivatized to yield 5-HT1A partial agonists
  • Pharmaceutical intermediate
Catalog No. 50-004-34096 Supplier Medchemexpress LLC Supplier No. HY763665G
(S)-N-Boc-3-hydroxypyrrolidine is a pharmaceutical intermediate. It can be used for the synthesis of other compounds and can be derivatized to yield 5-HT1A partial agonists.
  • Used for the synthesis of other compounds
  • Can be derivatized to yield 5-HT1A partial agonists
  • Pharmaceutical intermediate